Learn More
ILEX LIFE SCIENCES LLC SCHNEIDERS P04-90500 6 X 500ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0490500X6
Encompass_Preferred
PAN-Biotech Schneider’s Drosophila Medium, w/o: L-Glutamine, w: 0.4 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 0.4 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: Without glutamine. PHENOL RED: Without Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Originally developed for the culture of Drosophila cells, this medium is also suitable for the culture of other dipteran cell lines.
Catalog No. 50-273-3759