Learn More
Cayman Chemical Schsndra chns Gomsn D 1mg
SDP
Supplier:  Cayman Chemical 306971 MG
Encompass_Preferred
A lignan with diverse biological activities; an AChE inhibitor (IC50 = 7.84 µM); scavenges ABTS and DPPH radicals (IC50s = 24.5 and 203 µM, respectively); inhibits UGT1A1 and UGT1A3 by 89.9 and 83.1%, respectively, at 100 µM; selective for UGT1A1 and UGT1A3 over other UGT isoforms
Catalog No. 50-298-8397