Learn More
Electron Microscopy Sciences Science Markers, 6/PK
SDP
Supplier:  Electron Microscopy Sciences 62043G
Encompass_Preferred
Fine tip cryogenic markers are alcohol-resistant on most surfaces. Preliminary testing is required. Not suitable for exposure to acid/ethanol mixes. Color: Green.
Catalog No. NC1878882