Learn More
Med Vet International Operating Scissors, 6", Straight, Sharp/Sharp Tip, German Grade, Von Klaus
Supplier:  Med Vet International VK1030415
Encompass_Preferred
6" German Grade Straight Operating Scissors are a versatile surgical isntrument. These Surgical Scissors are curved and have a sharp/sharp tip and are used generally for cutting tougher tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6420