Learn More
Electron Microscopy Sciences Scissors with ESD Ceramic Tip, Style 450AZ, 115 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7832572
Encompass_Preferred
Silicone and Chloride Free. Recommended for soldering, high temperature (1400°C) and strong chemical environment applications. Stainless Steel Handles, ceramic tips.
Size: 115 mm
Material: Stainless steel
Catalog No. 50-193-1031