Learn More
Med Vet International Miltex Heath Suture Scissors, Serrated, German, 6.25in, 9-96
Supplier:  Med Vet International MIL996
Encompass_Preferred
Primarily used for suture removal. The hook tip helps the blades slide under sutures for an easy and safe process. Professionally sterilized for practicality, ease, and safety. The blades have a serrated finish to prevent slips. 6.25in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-264-3504