Learn More
World Precision Instrument Iris Scissors, 11.5cm, Curved, German
SDP
Supplier:  World Precision Instrument 501759G
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-822-802