Learn More
World Precision Instrument Iris Scissors, 10cm, SuperCut, Straight
SDP
Supplier:  World Precision Instrument 14218
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-822-290
 Restricted Item