Learn More
World Precision Instrument Mayo Scissors, 17 cm, Curved, Tungsten Carbide
SDP
Supplier:  World Precision Instrument 500221
Encompass
Mayo scissors come various lengths. Their construction is extremely durable, and Mayo scissors are used to cut connective tissues, fascia and sutures, primarily. The tips are semi-blunt, and they are available in curved or straight styles.
  •  
Catalog No. 50-822-389