Learn More
World Precision Instrument McPherson-Vannas Spring Scissors, 7cm, Curved, 3mm Blade
SDP
Supplier:  World Precision Instrument 501233
Encompass
McPherson-Vannas scissors are a fine pair of surgical scissors commonly used in ophthalmic procedures, specifically for delicate dissection and excision of ocular tissues. They may also be used in ophthalmic surgeries for cataract removal, glaucoma, and corneal transplantation. Other surgical specialties, like neurosurgery, plastic surgery, and ear, nose, and throat (ENT) surgery, may use McPherson-Vannas spring scissors for delicate dissection and cutting of tissues. These scissors have fine, sharp blades that are either curved or straight. These short, fine blades allow for precise cuts of delicate structures. Withe the spring handle, they are designed for excellent control and visibility during surgery, while minimizing trauma to surrounding tissues. The spring handles offer a stable grip for the surgeon, and they can also be used by either left- or right-handed operators.
Catalog No. 50-822-577