Learn More
Med Vet International Miltex Metzenbaum Scissors, 7in, Curved
Supplier:  Med Vet International MIL5182
Encompass_Preferred
The Miltex 5-182 Metzenbaum Scissors are surgical scissors designed for cutting delicate tissue and blunt dissection. These scissors are made of German grade stainless steel and have a length of 7in..
Catalog No. 50-264-3524