Learn More
World Precision Instrument Metzenbaum Scissors, 17.75 cm, Curved, Tungsten Carbide
SDP
Supplier:  World Precision Instrument 501745
Encompass
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
Catalog No. 50-822-775