Learn More
Med Vet International Miltex Operating Scissors, Standard Pattern, Curved, Sharp/Sharp, German, 6in, 5-37
Supplier:  Med Vet International MIL537
Encompass_Preferred
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. Curved blades with sharp/sharp tips. 6in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-283-1361