Learn More
Med Vet International Miltex Operating Scissors, Tungsten Carbide, Straight, Sharp/Blunt, German, 5.75in, 5-16TC
Supplier:  Med Vet International MIL516TC
Encompass_Preferred
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. Serrated blade helps prevent tissue slippage. These scissors are used for general, multipurpose cutting and dissecting. Equipped with straight blades with sharp/blunt tips. Tungsten coated for increased durability. One serrated blade to prevent slipping. 5.75in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-264-3526