Learn More
Med Vet International Miltex Or Scissors, 5.5in., Curved With Sharp/Blunt Tip, Tungsten Carbide
Supplier:  Med Vet International MIL546TC
Encompass_Preferred
The Miltex 5-46TC Operating Scissors have a length of 5.5in.. These standard operating scissors have a curved blade with sharp/blunt tip. They are made of German grade stainless steel and Tungsten Carbide.
Catalog No. 50-283-2356