Learn More
Med Vet International Miltex Olsen-Hegar Needle Holder & Scissors, Smooth Tungsten Carbide, German, 4.75in, PM-2165
Supplier:  Med Vet International MILPM2165
Encompass_Preferred
A needle holder, also called needle driver, is a surgical instrument, similar to a hemostat, used by doctors and surgeons to hold a suturing needle for closing wounds during suturing and surgical procedures. Coated with tungsten carbide for increased durability. 4.75in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-252-2284