Learn More
Med Vet International Hi-Level Bandage Scissors 5.5in German
Supplier:  Med Vet International VK0130199
Encompass_Preferred
Use Alternate Sku: VK140-2084The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose. Sterilization method- Steam
Catalog No. 50-209-2739