Learn More
Med Vet International 7.5-in Lister Bandage Scissors
Supplier:  Med Vet International 2145S
Encompass_Preferred
Lister bandage scissors are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric, and occasionally umbilical cord. The jaws of the Lister bandage scissors are angled with the lower blade being slightly longer.
Catalog No. 50-222-3979