Learn More
Cayman Chemical SclerotIorIn 5mg
SDP
Supplier:  Cayman Chemical 894605 MG
Encompass_Preferred
A natural product that inhibits soybean LO-1 (IC50 = 4.2 µM), cholesterol ester transfer protein (IC50 = 19.4 µM), Grb2-Shc interaction (IC50 = 22 µM), and antagonizes endothelin receptors (IC50 = 114 and 152 µM for human ETA and ETB, respectively)
Catalog No. 50-289-6617