Learn More
Medchemexpress LLC HY-N0228 5mg Medchemexpress, Scoparone CAS:120-08-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02285MG
Encompass_Preferred
Medchemexpress, HY-N0228 5mg Scoparone CAS:120-08-1 Scoparone is isolated from Artemisia capillaris, has anticoagulant, vasorelaxant antioxidant, anti-inflammatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0237