Learn More
Apexbio Technology LLC Scopine 498-45-3 25mg
SDP
Supplier:  Apexbio Technology LLC B136425
Encompass_Preferred
Scopine (CAS 498-45-3) is a tropane alkaloid recognized as a metabolic product of anisodine While anisodine functions as an 1-adrenergic receptor agonist involved in the management of acute circulatory shock scopine itself serves predominantly as a pharmacologically relevant intermediate in the metabolic pathway of these compounds In biomedical research scopine is utilized to investigate the metabolic fate and biotransformation processes of tropane alkaloids supporting studies focused on drug metabolism pharmacokinetics and the development of adrenergic receptor-targeting therapeutics
Catalog No. 50-000-92466