Learn More
Medchemexpress LLC Scopolamine hydrobromide trihydrate | 6533-68-2 | 99.86% | 438.31 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01089210MM/1ML
Encompass_Preferred
Scopolamine (Hyoscine) hydrobromide trihydrate is a high-affinity muscarinic antagonist that can cross the blood-brain barrier. It competitively antagonizes 5-HT3 receptors with an IC50 of 2.09 μM. This compound induces cognitive and memory deficits in animals and is used in research for preventing postoperative nausea and vomiting, motion sickness, and nervous system diseases.
  • High-affinity muscarinic antagonist
  • Can cross the blood-brain barrier
  • Competitively antagonizes 5-HT3 receptors with an IC50 of 2.09 μM
  • Induces cognitive and memory deficits in animals
  • Used for preventing postoperative nausea and vomiting
  • Used for motion sickness research
  • Used for nervous system diseases research
  • For research use only
  • Can be used to induce memory impairment models
Catalog No. 50-004-51760