Learn More
Medchemexpress LLC Scopolamine hydrobromide trihydrate | 6533-68-2 | MFCD00150066 | 99.9% | 438.31 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW010892100MG
Encompass_Preferred
Scopolamine hydrobromide trihydrate is a high-affinity muscarinic antagonist that can cross the blood-brain barrier. It acts as a competitive antagonist of 5-HT3 receptors. This compound can induce cognitive and memory deficits in animals and is used in research.
  • High-affinity muscarinic antagonist
  • Crosses the blood-brain barrier
  • Competitive antagonist of 5-HT3 receptors
  • Induces cognitive and memory deficits in animals
  • Used in research for preventing postoperative nausea and vomiting
  • Used in research for motion sickness
  • Used in research for nervous system diseases
Catalog No. 50-003-66337