Learn More
Perkin Elmer US LLC Scott/Cross-Flow Sample Introduction Torch Module for Avio 500
Supplier:  Perkin Elmer US LLC N0810606
Encompass_Preferred
This module kit includes: Outer Etched Scott Spray Chamber Component (N0791499) Inner Etched Scott Spray Chamber Component (N0582258) Spray Chamber External Drain (N0790613) GemTip Cross-Flow Nebulizer End Cap Assembly (N0780546) 1-Slot Quartz Torch (N0790131) Scott/Cross-Flow Adjustable Injector Adapter (N0790156) Spray Chamber Adapter (N0790296) 2.0 mm I.D. Alumina Injector (N0791183) Adjustable Torch Body Assembly (N0810128)
Catalog No. 50-211-2628