Learn More
Medchemexpress LLC SCP1-IN-2 | 2764615-56-5 | 98.77% | C19H17F3N2O6S2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1426831MG
Encompass_Preferred
SCP1-IN-2 (Compound SH T-65) is a potent and selective covalent inhibitor against SCP1. It promotes REST degradation and reduces transcriptional activity. High levels of REST protein drive tumor growth in some glioblastoma cells. SCP1-IN-2 has potential for research into glioblastoma where growth is driven by REST transcription activity.
  • Potent and selective covalent inhibitor against SCP1
  • Promotes REST degradation
  • Reduces transcriptional activity
  • Useful for glioblastoma research where growth is driven by REST transcription activity
Catalog No. 50-003-54118