Learn More
Medchemexpress LLC Scr7 pyrazine | 14892-97-8 | MFCD02167478 | 99.9% | 332.38 g·mol⁻¹ | C18H12N4OS | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10784525MG
Encompass_Preferred
SCR7 pyrazine is a small-molecule inhibitor of DNA ligase IV that blocks nonhomologous end-joining (NHEJ) and increases homology-directed repair (HDR) efficiency in genome-editing applications. It is supplied for research use only and is characterized by a defined molecular formula and molecular weight.
  • Inhibits DNA ligase IV and blocks NHEJ.
  • Enhances CRISPR/Cas9-mediated homology-directed repair.
  • High purity (~99.9%) suitable for research applications.
  • Supplied as solid and as a ready-to-use DMSO solution.
  • Stable when stored under recommended low-temperature conditions.
Catalog No. 50-002-42369