Learn More
BIOHIPPO Scramble-sh RFP Blasticidin
SDP
Supplier:  BIOHIPPO BHV194001801
Encompass
Scr-sh-RFP-BSD shRNA lentiviral vector expressing RFP with Blasticidin selection for studies in Each vial provides a nominal titer of 2x10E6 transducing units TU Scrambled shRNA Control Lentivirus High-titer shRNA lentiviral particles generated from a mixture of two independent scrambled shRNA control constructs The scrambled shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Recommended for use as a control with any of our shRNA or miRNA lentiviral particles
Catalog No. 50-003-31606