Learn More
CPC Scientific H-Phe-Ser-Leu-Leu-Arg-Asn-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific THRO006B
Encompass
This is a negative control peptide containing the reversed sequence of the first two amino acids of the receptor-activating (tethered ligand) peptide SFLLRN-NH2. It shows no detectable affinity for the binding sites. Whereas SFLLRN-NH2 effectively displaces [3H]haTRAP, thrombin receptor-activating peptide, the same peptide with serine and phenylalanine reversed, FSLLRN-NH2, does not.SEQUENCE: H-Phe-Ser-Leu-Leu-Arg-Asn-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: FSLLRN-NH2MOLECULAR FORMULA: C34H57N11O8MOLECULAR WEIGHT: 747.89STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [374898-11-0]SYNONYMS: (Phe,Ser)-Thrombin Receptor Activator Peptide 6, (Phe,Ser)-Thrombin Receptor 1 (1-6) (human), (Phe,Ser)-PAR-1 (1-6) (human), (Phe,Ser)-Proteinase Activated Receptor 1 (1-6) (human), FSLLRN, (Phe,Ser)-Coagulation Factor II Receptor (1-6) (human)RESEARCH AREA: Hematology, CardiovascularREFERENCES:Ahn, HS. et al. Mol. Pharmacol. 51, 350 (1997)SKU(s): THRO-006A,
Catalog No. 50-211-0569