Learn More
Essendant SCREENGRIDDLE10/20S
Supplier:  Essendant MMM20507
Encompass_Preferred
Designed to clean griddles that have been cooled down Tough aluminum oxide abrasive particles provide heavy-duty cleaning and removal of carbonized grease and food buildup on flat top griddles Replaces scrapers grill bricks and grill stones that disintegrate Use with Scotch-Brite Griddle Cleaning System handle and pad Scouring Pads/Sticks Type- Screen Material(s)- Fabric Backing Mineral Resin Size (WxL)- 4x5 5 Color(s)- Gray
Catalog No. 50-000-33029