Learn More
METTLER TOLEDO 3P INDUSTRIAL Screw Cap, 5/10 ml, 228 per pack
Supplier:  METTLER TOLEDO 3P INDUSTRIAL 30407436
Encompass_Preferred
A precision analytical balance with a 120g capacity and 0.1mg readability. It has fully automatic internal calibration and an RS-232 interface for connection to a computer or printer. The balance features a removable draft shield and level indicator. It has a stainless steel pan size of 3.5" diameter. This balance is designed for accurate weighing in laboratories, universities, industrial and manufacturing facilities. It has multiple weighing units including grams, milligrams, carats, and more. The balance provides reliable and repeatable results for precise weighing applications.
Catalog No. NC1506727