Learn More
CHEMSCIENCE Screw Cap and Bonded Septa, Size 9-425, Red PTFE/White Silicone 1mm(0.040),CS/1000
Supplier:  CHEMSCIENCE CSCS09BS
Encompass_Preferred
Screw Cap and Bonded Septa, Size 9-425, Red PTFE/White Silicone 1mm(0.040), PK/100,CS/10PKS
Catalog No. NC2681279