Learn More
Electron Microscopy Sciences Screw Cap Centrifuge Tube 50 ml, Co-Polymer, Sterile
SDP
Supplier:  Electron Microscopy Sciences 6476010
Encompass_Preferred
Premium quality general purpose centrifuge tubes with printed graduations. Polypropylene copolymer for excellent chemical resistance, withstanding tempserature of -40C to 125C; high centrifugation of 6000RCF. Exceptional clarity.
Material: Co-Polymer
Sterile: Sterile
Catalog No. 50-243-53