Learn More
Sarstedt Inc Screw cap tube, 30 ml, (LxØ): 107 x 25 mm, PP, 500/cs
Supplier:  Sarstedt Inc 60.545
Encompass
Screw cap tube, working volume: 30 ml, (LxØ): 107 x 25 mm, material: PP, conical base, transparent, screw cap, natural, cap assembled, sterile, 50 piece(s)/bag, 10 bags/case
Catalog No. NC3343311