Learn More
Perkin Elmer US LLC Venting Screw
Supplier:  Perkin Elmer US LLC N3141021
Encompass_Preferred
Screw for safe and convenient closing and venting of reaction vessels. Consists of a PEEK body with PTFE-TFM sealing bolt and O-Ring 2x1 FPM80.
Catalog No. 50-904-4500