Learn More
Chemglass Life Sciences GL-14 Screw Thread Tube, on 12mm OD x 1.5mm W tubing, 100mm OAL
SDP
Supplier:  Chemglass Life Sciences CG19401
Encompass_Preferred
GL-14 Screw Thread Tube, on 12mm OD x 1.5mm W tubing, 100mm OAL. Screw thread tube for replacement on apparatus using the GL thread, which is manufactured to I.S.O. specifications. (GL is the I.S.O. designation for GLASS.) To be used with a CG-196 solid cap with liner or a CG-195 cap with aperture and CG-197 silicone sealing ring.
Catalog No. CG19401