Learn More
Uline Screw Top Pail - 1.25 Gallon, White Lid | 5/case
Supplier:  Uline S18114WCS
Encompass_Preferred
Quickly open and reseal for easy access. Vacuum tight ¿ Use with granulars, fine powders. Tamper evident with use of cable ties. Lid includes neoprene gasket for moisture-resistant storage. Molded, high-density polyethylene. FDA compliant.
Catalog No. NC1874167