Learn More
Cayman Chemical Scriptaid Inhibitors
SDP
Supplier:  Cayman Chemical 105721 MG
Encompass_Preferred
An HDAC inhibitor that has an optimal effective concentration of 6-8 μM in a cell-based assay, is less toxic than trichostatin A, and works in a wide variety of biological systems; induces cell cycle arrest in cancer cells in vitro and in vivo; facilitates the cloning of inbred mouse strains produced by somatic cell nuclear transfer
Catalog No. 50-288-7282