Learn More
Aviva Systems Biology Corp SCRT2 Antibody - middle region (ARP50735_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50735_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCRT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SCRT2. Alternate names: ZNF898BApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rat, Yeast
Catalog No. 50-014-60421