Learn More
Medchemexpress LLC Scutellarin methyl ester | 119262-68-9 | 5 MG

Supplier: Medchemexpress LLC HYN69255MG
Scutellarin methyl ester is a constituent of Breviscapine, a crude extract composed of various flavonoids derived from Erigeron breviscapus. It is intended for research use only.
- Purity: 98.17%
- Molecular weight: 476.39
- Formula: C22H20O12
- Appearance: Solid
- Color: Light yellow to yellow
- Structure classification: Flavonoids, flavones, phenols, polyphenols
- Initial source: Plants other families
- Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
- Shipping: Room temperature in continental US; may vary elsewhere.
- Solubility: In vitro: DMSO : 25 mg/mL (52.48 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.