Learn More
Waters Corp BioResolve SCX mAb Start Up Kit, 2.1 x 50 mm
Supplier: Waters Corp 176004355
This start up kit is designed for reproducible, high-resolution, charge-based separations of monoclonal antibodies and other proteins in LC and LC-MS applications. It includes a 3 μm, corrosion-resistant column with non-porous, polymer-based particles and negatively charged sulfonic acid ligands. The kit also provides an mAb Charge Variant Standard, pH Concentrates, and two 1000 mL containers.
- Utilizes 3 μm, corrosion-resistant columns for LC and LC-MS applications.
- Provides reproducible, high-resolution, charge-based separations for mAbs and other proteins.
- Engineered for superior component recovery, peak shape, and resolution.
- Compatible with UHPLC, HPLC, and UPLC systems.
- Includes mAb Charge Variant Standard and pH Concentrates.
- Comes with two Certified LDPE 1000 mL containers.
- Recommended maximum pressure of 10,000 psi.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.