Learn More
Cayman Chemical scyloInosItol hekIsphsph 10mg
SDP
Supplier:  Cayman Chemical 900233910 MG
Encompass_Preferred
A stereoisomer of IP6; identified in soil samples from Madagascar, New Zealand, and the Falkland Islands and in freshwater sediment from Danish lakes; has been used as a negative control for the stereospecific effects of myo-inositol hexakisphosphate on IK,in in intact guard cells
Catalog No. 50-289-3212