Learn More
iQ Biosciences SD Rat Splenocytes, 25M cells/vial
Supplier:  iQ Biosciences IQBRSP101
Encompass
High-quality, high-viability (typically > 70%) splenocytes from the Sprague Dawley rat, a commonly-used outbred rat strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2855624