Learn More
Aat Bioquest SDA alkyne, 1mg .
SDP
Supplier:  Aat Bioquest 39005
Encompass_Preferred
SDA alkyne contains both diazirine photoaffinity moiety and alkyne functional group. It is an excellent building block to introduce photoaffinity function into an azide-modified biomolecule via the well-known click chemistry. Diazirines are known for their ability to undergo photochemical reactions when exposed to ultraviolet (UV) light, specifically by forming highly reactive carbene intermediates that react with nearby molecules, forming covalent bonds. This property makes diazirines useful for studying protein-protein, protein-nucleic acid interactions, ligand-receptor binding, and other biomolecular interactions. It's important to note that diazirines are highly reactive and can be challenging to handle due to their instability. They require careful storage and handling, typically in a controlled environment.
Catalog No. NC3930624