Learn More
MOLTOX Molecular Toxicology Inc Sabouraud Dextrose Agar (SDA) w/Lecithin & Tween 80, pH 5.6 +/- 0.2, 100 mL/bottle, Store at Room Temperature
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 26740
Encompass
Sabouraud Dextrose Agar is used for the cultivation of pathogenic and nonpathogenic fungi such as dermatophytes and can be more selective with the addition of antimicrobics. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. NC1005590