Learn More
Alkali Scientific SDS, 20% Solution [Sodium dodecylsulfate 20% Solution][Lauryl sulfate 20% Solution], 5 L
SDP
Supplier:  Alkali Scientific SDS007CP
Encompass_Preferred
Sodium Dodecyl Sulfate (SDS) 20% Solution in a 5L container is ideal for large-scale laboratory applications requiring high quantities of detergent. The solution is commonly used in molecular biology and biochemistry laboratories for protein electrophoresis, cell lysis, and membrane solubilization. With a concentration of 20%, SDS efficiently disrupts cellular structures by denaturing proteins and membranes, enabling the extraction of proteins for subsequent analysis. This large format provides a cost-effective solution for labs that require significant amounts of SDS for routine protocols, including SDS-PAGE, protein purification, and analytical testing of various biochemical samples.
Catalog No. 50-006-61236