Learn More
AB Sciex LLC SDS-MW Gel Multi Buffer Pack (4 bottle pack of SDS-MW gel)
Supplier:  AB Sciex LLC A30341
Encompass
SDS-MW Gel Buffer is a 4-pack of the SDS-MW gel used in the SDS-MW Kit (Part Number 390953) and the IgG Purity and Heterogeneity Kit assay (Part Number a10663). Each multi pack contains 4 individual bottles of buffer.
Catalog No. NC9899107
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.