Learn More
IBI SCIENTIFIC 3P Sodium Dodecyl Sulfate | 151-21-3 | MFCD00036175 | 500 g
Supplier:  IBI SCIENTIFIC 3P IB07062
Encompass_Preferred
Sodium Dodecyl Sulfate (SDS) - 500g. Ionic detergent used to denature proteins in hybridization, nucleic acid purification and electrophoresis buffer systems. Disrupts cell membranes via dissociation of nucleic acid protein complexes.
Catalog No. NC9983815