Learn More
Boston Bioproducts Inc Laemmli SDS-Sample Buffer (4X, Non-Reducing) - 50ml
Supplier:  Boston Bioproducts Inc BP110NR50ML
Encompass
The 4X non-reducing laemmli sample buffer is a critical component in the preparation of protein samples for SDS-PAGE, ensuring resolution of protein bands. Its composition typically includes Tris-HCl, glycerol, SDS, and a tracking dye like Bromophenol Blue, which provide a suitable environment for protein electrophoresis. Non-reducing Laemmli Sample Buffers do not contain reducing agents, preserving disulfide bonds in proteins.
Catalog No. NC9506238