Learn More
Medchemexpress LLC HY-13059A 10mg Medchemexpress, SDZ 220-581 (Ammonium salt) CAS:179411-94-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13059A10MG
Encompass_Preferred
Medchemexpress, HY-13059A 10mg SDZ 220-581 (Ammonium salt) CAS:179411-94-0 SDZ 220-581 Ammonium salt is an oral bioavailability, potent, competitive NMDA receptor antagonist with pK i value of 7.7 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2650