Learn More
Medchemexpress LLC HY-108938 5mg Medchemexpress, SDZ285428 CAS:174262-13-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1089385MG
Encompass_Preferred
Medchemexpress, HY-108938 5mg SDZ285428 CAS:174262-13-6 SDZ285428 is a CYP51 inhibitor. SDZ285428 inhibits Trypanosoma cruzi (TC) CYP51 with I/E2 <1 (5 min) and I/E2=9 (1 h). SDZ285428 inhibits Trypanosoma brucei (TB) CYP51 with I/E2 <1 (5 min) and I/E2=35 (1 h). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0755